Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

177881

Sigma-Aldrich

Tris(hydroxymethyl)phosphine

90%

Synonym(s):

Phosphinidynetrimethanol, Trimethylolphosphine, Tris(methanol)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)3P
CAS Number:
Molecular Weight:
124.08
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

bp

111-113 °C/2.5 mmHg (lit.)

mp

48-56 °C (lit.)

functional group

phosphine

SMILES string

OCP(CO)CO

InChI

1S/C3H9O3P/c4-1-7(2-5)3-6/h4-6H,1-3H2

InChI key

JMXMXKRNIYCNRV-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baobin Li et al.
Nature communications, 8(1), 666-666 (2017-09-25)
The O-linked β-N-acetyl glucosamine (O-GlcNAc) modification dynamically regulates the functions of numerous proteins. A single human enzyme O-linked β-N-acetyl glucosaminase (O-GlcNAcase or OGA) hydrolyzes this modification. To date, it remains largely unknown how OGA recognizes various substrates. Here we report
Christoph Krettler et al.
Analytical biochemistry, 474, 89-94 (2014-12-20)
Use of the reductant dithiothreitol (DTT) as a substrate for measuring vitamin K 2,3-epoxide reductase (VKOR) activity in vitro has been reported to be problematic because it enables side reactions involving the vitamin K1 2,3-epoxide (K1>O) substrate. Here we characterize
Rhushikesh A Kulkarni et al.
Nature chemical biology, 15(4), 391-400 (2019-02-06)
Hereditary cancer disorders often provide an important window into novel mechanisms supporting tumor growth. Understanding these mechanisms thus represents a vital goal. Toward this goal, here we report a chemoproteomic map of fumarate, a covalent oncometabolite whose accumulation marks the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service