Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

176524

Sigma-Aldrich

3-Bromofuran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3BrO
CAS Number:
Molecular Weight:
146.97
Beilstein:
105337
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

103 °C (lit.)

density

1.635 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Brc1ccoc1

InChI

1S/C4H3BrO/c5-4-1-2-6-3-4/h1-3H

InChI key

LXWLEQZDXOQZGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Bromofuran was used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam McCluskey et al.
Bioorganic chemistry, 31(1), 68-79 (2003-04-17)
Diels-Alder addition of furans (furan, furfuryl alcohol, and 3-bromofuran) to maelic anhydride yields three distinct 5,6-dehydronorcantharidins. Hydrogenation of (4,10-dioxatricyclo[5.2.1.0]decane-3,5-dione) (4a), in dry ethanol affords the monoester (7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylic aid monoethyl ester) (6). Subsequent transesterification affords a series of monoesters (7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid
Ann Rowley Kelly et al.
Journal of the American Chemical Society, 130(12), 4097-4104 (2008-03-05)
Furans and pyrroles are important synthons in chemical synthesis and are commonly found in natural products, pharmaceutical agents, and materials. Introduced herein are three methods to prepare 2-substituted 3-furfurals starting from 3-furfural, 3-bromofuran, and 3-vinylfurans. Addition of a variety of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service