Skip to Content
MilliporeSigma
All Photos(1)

Documents

176095

Sigma-Aldrich

2,2′-Oxydiethylamine dihydrochloride

97%

Synonym(s):

2,2′-Oxybis(ethylamine) dihydrochloride, 2-Aminoethyl ether dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(CH2CH2NH2)2 · 2HCl
CAS Number:
Molecular Weight:
177.07
Beilstein:
3672341
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

230-234 °C (lit.)

SMILES string

Cl.Cl.NCCOCCN

InChI

1S/C4H12N2O.2ClH/c5-1-3-7-4-2-6;;/h1-6H2;2*1H

InChI key

KTCUXFVANABSPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-Oxybis(ethylamine) dihydrochloride was used in the synthesis of macrocyclic bisimines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preparation of thiophene-based azacryptand Mannich bases.
Journal of the Chemical Society. Perkin Transactions 1, 6, 717-724 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service