Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

167703

Sigma-Aldrich

3-(Trifluoromethyl)benzyl chloride

97%

Synonym(s):

α′-Chloro-α,α,α-trifluoro-m-xylene, 3-(Chloromethyl)benzotrifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2Cl
CAS Number:
Molecular Weight:
194.58
Beilstein:
743242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.464 (lit.)

density

1.254 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cccc(CCl)c1

InChI

1S/C8H6ClF3/c9-5-6-2-1-3-7(4-6)8(10,11)12/h1-4H,5H2

InChI key

XGASTRVQNVVYIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Trifluoromethyl)benzyl chloride reacts with sodium salts of N,N-disubstituted dithiocarbamic acids to yield series of dithiocarbamates.

Application

3-(Trifluoromethyl)benzyl chloride was used in the synthesis of 4-nitro-3-trifluoromethyl-[N-(4-hydroxyphenyl)]benzamide.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of poly (arylene ether amide) s containing CF 3 groups by nitro displacement reaction of AB-type monomers.
Lee HS and Kim SY.
Macromolecular Rapid Communications, 23(12), 665-671 (2002)
Mehlika D Altıntop et al.
Archiv der Pharmazie, 346(8), 571-576 (2013-07-25)
In the present paper, a novel series of dithiocarbamates was synthesized via the treatment of 4-(trifluoromethyl)benzyl chloride with appropriate sodium salts of N,N-disubstituted dithiocarbamic acids. The chemical structures of the compounds were elucidated by (1) H NMR, mass spectral data
William L Scott et al.
Journal of combinatorial chemistry, 11(1), 14-33 (2008-12-25)
Distributed Drug Discovery (D(3)) proposes solving large drug discovery problems by breaking them into smaller units for processing at multiple sites. A key component of the synthetic and computational stages of D(3) is the global rehearsal of prospective reagents and
William L Scott et al.
Journal of combinatorial chemistry, 11(1), 34-43 (2008-12-25)
For the successful implementation of Distributed Drug Discovery (D(3)) (outlined in the accompanying Perspective), students, in the course of their educational laboratories, must be able to reproducibly make new, high quality, molecules with potential for biological activity. This article reports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service