All Photos(1)
About This Item
Linear Formula:
HOC6H3(OCH3)CN
CAS Number:
Molecular Weight:
149.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
mp
85-87 °C (lit.)
functional group
nitrile
SMILES string
COc1cc(ccc1O)C#N
InChI
1S/C8H7NO2/c1-11-8-4-6(5-9)2-3-7(8)10/h2-4,10H,1H3
InChI key
QJRWLNLUIAJTAD-UHFFFAOYSA-N
General description
4-Hydroxy-3-methoxybenzonitrile on treatment with sodium perborate yields 4-hydroxy-3-methoxybenzamide.
Application
4-Hydroxy-3-methoxybenzonitrile was used in the synthesis of 1,5-bis(4-cyano-2-methoxyphenoxy)-3-oxapentane.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dorota Maciejewska et al.
European journal of medicinal chemistry, 48, 164-173 (2011-12-28)
A series of 20 pentamidine analogs were prepared using 2 general Schemes that evaluated heteroatoms, sulfobenzene and alkanediamide groups in the aliphatic linker and methoxy substituents attached to the benzene rings for efficacy against the fungal pathogen, Pneumocystis carinii in
Jerzy W Jaroszewski et al.
Natural product research, 19(3), 291-294 (2005-02-11)
Extract of Naravelia zeylanica (Ranunculaceae) yielded three simple benzamides, 3,4-methylenedioxybenzamide, 4-methoxybenzamide and 4-hydroxy-3-methoxybenzamide. These simple C6C1 metabolites have been encountered as natural products for the first time. The compounds were identified by direct comparison of their spectral (1H- and 13C-NMR)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service