Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

161004

Sigma-Aldrich

1,3,5-Triisopropylbenzene

95%

Synonym(s):

1,3,5-Tri(propan-2-yl)benzene, 1,3,5-Tris(1-methylethyl)benzene, 1,3,5-Tris(isopropyl)benzene, 1,3,5-Tris(propan-2-yl)benzene, 2,4,6-Triisopropylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3[CH(CH3)2]3
CAS Number:
Molecular Weight:
204.35
Beilstein:
1862750
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

232-236 °C (lit.)

density

0.845 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cc(cc(c1)C(C)C)C(C)C

InChI

1S/C15H24/c1-10(2)13-7-14(11(3)4)9-15(8-13)12(5)6/h7-12H,1-6H3

InChI key

VUMCUSHVMYIRMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3,5-Triisopropylbenzene was employed as swelling agent during the synthesis of mesoporous silicas with two-dimensional hexagonal pores. It was also employed as micelle expander in the synthesis of highly ordered mesoporous SBA-15 silica and magnetic mesoporous silica nanoparticles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Cao et al.
Journal of colloid and interface science, 361(2), 472-476 (2011-06-28)
Highly ordered mesoporous SBA-15 silica with large pore diameter of 18 nm (nominal BJH pore diameter ~22 nm) and short pore length (~500 nm) was synthesized using a micelle expander 1,3,5-triisopropylbenzene in the absence of ammonium fluoride by employing short
Jixi Zhang et al.
Journal of colloid and interface science, 361(1), 16-24 (2011-06-22)
Magnetic mesoporous silica nanoparticles (M-MSNs) are emerging as one of the most appealing candidates for theranostic carriers. Herein, a simple synthesis method of M-MSNs with a single Fe(3)O(4) nanocrystal core and a mesoporous shell with radially aligned pores was elaborated
Selvedin Telalović et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(7), 2077-2088 (2011-01-25)
Bimetallic three-dimensional amorphous mesoporous materials, Al-Zr-TUD-1 materials, were synthesised by using a surfactant-free, one-pot procedure employing triethanolamine (TEA) as a complexing reagent. The amount of aluminium and zirconium was varied in order to study the effect of these metals on
Zunfang Hu et al.
Journal of environmental sciences (China), 25(1), 181-187 (2013-04-17)
Immobilization of enzymes on mesoporous silicas (MS) allows for good reusability. MS with two-dimensional hexagonal pores in diameter up to 14.13 nm were synthesized using Pluronic P123 as template and 1,3,5-triisopropylbenzene as a swelling agent in acetate buffer. The surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service