Skip to Content
MilliporeSigma
All Photos(1)

Documents

157147

Sigma-Aldrich

Valeroyl chloride

98%

Synonym(s):

Pentanoic acid chloride, n-Butanecarbonyl chloride, n-Pentanoyl chloride, n-Valeryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3COCl
CAS Number:
Molecular Weight:
120.58
Beilstein:
506293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.42 (lit.)

bp

125-127 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

SMILES string

CCCCC(Cl)=O

InChI

1S/C5H9ClO/c1-2-3-4-5(6)7/h2-4H2,1H3

InChI key

XGISHOFUAFNYQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Valeroyl chloride was used in the enantioselective synthesis of propylisopropyl acetamide (PID), an amide analog of the major antiepileptic drug valproic acid. It was also used in the synthesis of (4R, 5S-4-methyl-3-(1-oxopentyl)-5-phenyl-oxazolidinone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Hauck et al.
Pharmaceutical research, 9(7), 850-855 (1992-07-01)
The teratogenic activities of R(+)- and S(-)-2-n-propyl-4-pentynoic acid (R and S-4-yn-VPA), the enantiomers of the highly teratogenic valproic acid (VPA) analogues (+/-)-4-yn-VPA, were investigated in mice. The enantiomers were prepared via asymmetric synthesis, each in three steps employing the chiral
O Spiegelstein et al.
Chirality, 11(8), 645-650 (1999-09-01)
Propylisopropyl acetamide (PID), an amide analogue of the major antiepileptic drug valproic acid (VPA), possesses favorable anticonvulsant and CNS properties. PID contains one chiral carbon atom and therefore exists in two enantiomeric forms. The purpose of this work was to
Janclei P Coutinho et al.
Talanta, 134, 256-263 (2015-01-27)
In this work a multivariate statistical tool (Derringer and Suich optimization) was proposed for the separation of seventeen capsinoids (natural and synthetic) using the UHPLC-DAD chromatography. Capsinoids were analyzed at 280 nm. The variables optimized were the mobile phase (water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service