Skip to Content
MilliporeSigma
All Photos(1)

Documents

140295

Sigma-Aldrich

Decanoyl chloride

98%

Synonym(s):

Capric acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COCl
CAS Number:
Molecular Weight:
190.71
Beilstein:
507055
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.57 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

94-96 °C/5 mmHg (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCC(Cl)=O

InChI

1S/C10H19ClO/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3

InChI key

IPIVAXLHTVNRBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Decanoyl chloride enhances the activity of proleather from Bacillus sp. during the synthesis of poly(lactic acid) in organic solvents.

Decanoyl chloride is used as a reactive acylating agent in organic synthesis. It is also employed in the formation of amides and esters.

Application

Decanoyl chloride was used in the synthesis of oligomers composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

284.0 °F

Flash Point(C)

140 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Berl et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 7(13), 2798-2809 (2001-08-07)
Molecular strands composed of alternating 2,6-diaminopyridine and 2,6-pyridinedicarbonyl units have been designed to self-organize into single stranded helical structures upon forming intramolecular hydrogen bonds. Pentameric strands 11, 12, and 14, heptameric strands 1 and 20, and undecameric strand 15 have
Multivariate CuBTC metal--organic framework with enhanced selectivity, stability, compatibility, and processability
Peterson GW, et al.
Chemistry of Materials, 31(20), 8459-8465 (2019)
Kelley A Distel et al.
Bioresource technology, 96(5), 617-623 (2004-10-27)
Proleather from Bacillus sp. was chemically modified with decanoyl chloride for enhanced activity for the preparation of poly(lactic acid) in organic solvents. The modified enzyme was highly soluble (up to 44 mg-protein/ml) and active in various organic solvents including chloroform
Dalia S Abdelhamid et al.
Biomaterials, 53, 32-39 (2015-04-22)
Cardiovascular disease initiates with the atherogenic cascade of scavenger receptor- (SR-) mediated oxidized low-density lipoprotein (oxLDL) uptake. Resulting foam cell formation leads to lipid-rich lesions within arteries. We designed amphiphilic macromolecules (AMs) to inhibit these processes by competitively blocking oxLDL
Janclei P Coutinho et al.
Talanta, 134, 256-263 (2015-01-27)
In this work a multivariate statistical tool (Derringer and Suich optimization) was proposed for the separation of seventeen capsinoids (natural and synthetic) using the UHPLC-DAD chromatography. Capsinoids were analyzed at 280 nm. The variables optimized were the mobile phase (water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service