Skip to Content
MilliporeSigma
All Photos(1)

Documents

139327

Sigma-Aldrich

3-Nitrobenzamidine hydrochloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
201.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

impurities

5% ammonium chloride

mp

245-248 °C (lit.)

SMILES string

Cl.NC(=N)c1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7N3O2.ClH/c8-7(9)5-2-1-3-6(4-5)10(11)12;/h1-4H,(H3,8,9);1H

InChI key

DKNQVJWYIUJWNC-UHFFFAOYSA-N

Application

3-Nitrobenzamidine hydrochloride was used as reagent during the synthesis of (Z)-3-benzoylamino-4-dimethylamino-2-oxo-3-butene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformations of (Z)-2-benzoylamino-4-dimethylamino-2-oxo-3-butene and (E)-3-benzoylamino-4-cyano-2-oxo-3-butene into pyrimidine, pyrazole and isoxazole derivatives.
Bratusek U, et al.
ARKIVOC (Gainesville, FL, United States), 77, 86-86 (2003)
J A Raleigh et al.
Cancer clinical trials, 3(1), 55-62 (1980-01-01)
Normal tissue toxicity of nitroaromatic radiosensitizers may originate in radiosensitizer/nitroreductase interaction. A study of two mammalian cell nitroreductases, xanthine oxidase and NADH cytochrome c reductase, shows that the efficiency of electron transfer is dependent on sensitizer electron affinity and not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service