Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

136603

Sigma-Aldrich

1-Ethyl-2-methylindole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N
CAS Number:
Molecular Weight:
159.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.587 (lit.)

bp

266-267 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCn1c(C)cc2ccccc12

InChI

1S/C11H13N/c1-3-12-9(2)8-10-6-4-5-7-11(10)12/h4-8H,3H2,1-2H3

InChI key

XMOWAIVXKJWQBJ-UHFFFAOYSA-N

Application

1-Ethyl-2-methylindole may be used in synthesis of hetrocyclic indole compounds.
Reactant for preparation of:
  • (2-hydroxyethyl)ammonium salts of 1-R-indol-3-ylsulfanyl(sulfonyl)alkanecarboxylic acids as potential immunomodulators
  • Three-bladed propeller-shaped molecules
  • Potential non-steroidal aromatase inhibitors
  • Antimycobacterial agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service