Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

127663

Sigma-Aldrich

3-Nitrobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCl
CAS Number:
Molecular Weight:
185.56
Beilstein:
777186
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

275-278 °C (lit.)

mp

31-34 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(c1)C(Cl)=O

InChI

1S/C7H4ClNO3/c8-7(10)5-2-1-3-6(4-5)9(11)12/h1-4H

InChI key

NXTNASSYJUXJDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitrobenzoyl chloride was used in the synthesis of 3-aminobenzamide. It was also used to synthesize potential carbocyclic minor groove binders.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>233.6 °F

Flash Point(C)

> 112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danuta Bartulewicz et al.
Farmaco (Societa chimica italiana : 1989), 59(3), 211-214 (2004-02-28)
New carbocyclic potential minor groove binders were synthesised, using 3-nitrobenzoyl chloride and aliphatic alpha,omega-diamines with three, four and five methylene fragments. The half structures, compounds IV-VI can be compared to bis-amidines, compounds X-XII to bis-netropsin. All of the compounds were
Meng-Ge Huangfu et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-03-07)
The rapid development of advanced high-frequency mobile communication techniques has advanced urgent requirements for polymer materials with high-temperature resistance and good dielectric properties, including low dielectric constants (low-Dk) and low dielectric dissipation factors (low-Df). The relatively poor dielectric properties of
B W Durkacz et al.
European journal of biochemistry, 121(1), 65-69 (1981-12-01)
DNA damage and repair was assayed by the loss and restoration of DNA supercoiling in nucleoids. This technique was used to assess the effects of inhibition of (ADP-ribose)n biosynthesis by 3-aminobenzamide on the capacity to repair DNA of mouse leukaemia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service