Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

127493

Sigma-Aldrich

4-Methylpyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Beilstein:
105781
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

141-142 °C (lit.)

density

1.031 g/mL at 25 °C (lit.)

SMILES string

Cc1ccncn1

InChI

1S/C5H6N2/c1-5-2-3-6-4-7-5/h2-4H,1H3

InChI key

LVILGAOSPDLNRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylpyrimidine was used in the synthesis of crystalline trans-[RuCl(3)(NO)L(2)] complexes. 4-Methylpyrimidine was also used to bridge dinuclear oxovanadium(IV) complexes.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosuke Kitase et al.
Dalton transactions (Cambridge, England : 2003), 49(35), 12210-12214 (2020-07-02)
A Hofmann-type two-step spin-crossover (SCO) complex with a pyrimidine derivative ligand, Fe(4-methylpyrimidine)2[Au(CN)2]2, was synthesized, and this complex shows a two-step SCO phenomenon in the intermediate state. Symmetry breaking also occurs in the intermediate state. These results reveal three spin states
T Ishida et al.
Inorganic chemistry, 40(27), 7059-7064 (2002-01-05)
Dinuclear oxovanadium(IV) complexes bridged by pyrimidine derivatives, L[VO(hfac)(2)](2) [L = pyrimidine (PM), 4-methylpyrimidine (MPM), 4,6-dimethylpyrimidine, 4-aminopyrimidine, and quinazoline; hfac = 1,1,1,5,5,5-hexafluoropentane-2,4-dionate], were synthesized and characterized. All of them showed intramolecular ferromagnetic interaction, and the magnetic susceptibilities were analyzed on the
Oleg O Glebov et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(12), 4830-4836 (2015-03-27)
Membrane trafficking of AMPA receptors (AMPARs) is critical for neuronal function and plasticity. Although rapid forms of AMPAR internalization during long-term depression (LTD) require clathrin and dynamin, the mechanisms governing constitutive AMPAR turnover and internalization of AMPARs during slow homeostatic
Gabriella Tamasi et al.
Journal of inorganic biochemistry, 102(10), 1874-1884 (2008-08-08)
The reaction of RuCl(3)(NO).H(2)O with 4-methylpyrimidine (MePYM) and ethylisonicotinate (EINT), in absolute ethanol at 40-55 degrees C afforded crystalline trans-[RuCl(3)(NO)L(2)] complexes. Structural studies via X-ray diffraction, and spectroscopic methods (NMR, IR, UV-visible (UV-Vis)) revealed that the molecular structures have the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service