Skip to Content
MilliporeSigma
All Photos(2)

Documents

117501

Sigma-Aldrich

2,6-Dinitrobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3CHO
CAS Number:
Molecular Weight:
196.12
Beilstein:
2113951
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

120-122 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1c(cccc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C7H4N2O5/c10-4-5-6(8(11)12)2-1-3-7(5)9(13)14/h1-4H

InChI key

WHFZQNNDIJKLIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dinitrobenzaldehyde is the major intermediate formed by the combined ozonation of 2,6-dinitrotoluene with hydrogen peroxide and UV radiation.

Application

2,6-Dinitrobenzaldehyde was used in the synthesis of free base tetrakis(2,6-dinitrophenyl) porphyrin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mori et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(7), 731-741 (1989-07-01)
1. Unchanged 2,6-dinitrotoluene (2,6-DNT), 2-amino-6-nitrotoluene, 2,6-dinitrobenzyl alcohol, 2-amino-6-nitrobenzyl alcohol, conjugated 2,6-dinitrobenzyl alcohol and conjugated 2-amino-6-nitrobenzyl alcohol were detected in urine of male Wistar rats dosed with 2,6-DNT. The major metabolite was conjugated 2,6-dinitrobenzyl alcohol, which accounted for about 1.5% of
Synthesis and spectroscopic characterization of bis-pocket porphyrins: tetrakis (2', 6'-dinitrophenyl) porphyrin and catalytic activity of a manganese (III) chloride derivative in alkane oxidation.
Quintana CA, et al.
Inorganic Chemistry, 28(18), 3421-3425 (1989)
Nitroaromatic hydrocarbon ozonation in water. 2. Combined ozonation with hydrogen peroxide or UV radiation.
Beltran FJ, et al
Industrial & Engineering Chemistry Research, 37(1), 32-40 (1998)
M Sayama et al.
Mutation research, 243(1), 47-52 (1990-01-01)
The mutagenicities and theoretical reactivity indices of 2,4-dinitrobenzaldehyde (2,4-DNBAl) and 2,6-dinitrobenzaldehyde (2,6-DNBAl) were investigated using Salmonella typhimurium strains TA98, TA98NR, TA98/1,8-DNP6, and TA100, TA100NR and TA100/1,8-DNP6, by means of the modified intermediate neglect of differential overlap/3 (MINDO)/3) method. The mutagenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service