Skip to Content
MilliporeSigma
All Photos(2)

Documents

109746

Sigma-Aldrich

1-Chloroheptane

99%

Synonym(s):

Heptyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6Cl
CAS Number:
Molecular Weight:
134.65
Beilstein:
1697159
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.425 (lit.)

bp

159-161 °C (lit.)

mp

−69 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCl

InChI

1S/C7H15Cl/c1-2-3-4-5-6-7-8/h2-7H2,1H3

InChI key

DZMDPHNGKBEVRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Chloroheptane was used in the coupling of a GC to the ICP-MHMS-FPC for the determination of halogenated hydrocarbons and organometallic species.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barnes JH, et al.
Journal of Analytical Atomic Spectrometry, 19(6), 751-756 (2004)
Thomas Bielewicz et al.
Nanotechnology, 27(35), 355602-355602 (2016-07-28)
Two-dimensional colloidal nanosheets represent very attractive optoelectronic materials. They combine good lateral conductivity with solution-processability and geometry-tunable electronic properties. In the case of PbS nanosheets, so far synthesis has been driven by the addition of chloroalkanes as coligands. Here, we
Jonas Winters et al.
Physical chemistry chemical physics : PCCP, 21(7), 4053-4062 (2019-02-05)
Polyaramids are a class of high-performance polymers, known for their high mechanical strength and chemical and thermal stability. Their ability to create a network of intermolecular hydrogen bonds causes them to be very poorly soluble in conventional solvents. Hazardous solvents
Maxwell Wei-Hao Li et al.
Journal of chromatography. A, 1620, 461002-461002 (2020-03-08)
Micro gas chromatography (µGC) is a technique developed for rapid, in situ analysis of volatile organic compounds (VOCs) for environmental protection, industrial monitoring, and toxicology. While reduced µGC size and power requirements allow for increased portability, the low moisture and
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service