Skip to Content
MilliporeSigma
All Photos(2)

Documents

105880

Sigma-Aldrich

Diphenyl phthalate

99%

Synonym(s):

Phenyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C6H5)2
CAS Number:
Molecular Weight:
318.32
Beilstein:
2473390
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

74-76 °C (lit.)

SMILES string

O=C(Oc1ccccc1)c2ccccc2C(=O)Oc3ccccc3

InChI

1S/C20H14O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h1-14H

InChI key

DWNAQMUDCDVSLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimitra Diamantidou et al.
Journal of chromatography. A, 1603, 165-178 (2019-06-27)
An Ultra High Performance Liquid Chromatography - Tandem Mass Spectrometry method has been developed for the analysis of 12 phthalate esters in Greek grape marc spirits. The phthalates were separated on a U-VDSpher PUR 100 C18-E (100 mm x 2.0 mm, 1.8 μm) column by
Wei Jia et al.
Journal of chromatography. A, 1362, 110-118 (2014-08-27)
A new analytical method was developed and validated for simultaneous analysis of 27 phthalates in milk and milk products. Response surface methodology was employed to optimize a quick, easy, cheap, effective, rugged, and safe (QuEChERS) sample preparation method. Ultrahigh-performance liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service