Merck
Heterozygous deletion of sarcolipin maintains normal cardiac function.
American journal of physiology. Heart and circulatory physiology (2015-11-01)
Daisuke Shimura, Yoichiro Kusakari, Tetsuo Sasano, Yasuhiro Nakashima, Gaku Nakai, Qibin Jiao, Meihua Jin, Tomohiro Yokota, Yoshihiro Ishikawa, Atsushi Nakano, Nobuhito Goda, Susumu Minamisawa
RESUMEN
Sarcolipin (SLN) is a small proteolipid and a regulator of sarco(endo)plasmic reticulum Ca(2+)-ATPase. In heart tissue, SLN is exclusively expressed in the atrium. Previously, we inserted Cre recombinase into the endogenous SLN locus by homologous recombination and succeeded in generating SLN-Cre knockin (Sln(Cre/+)) mice. This Sln(Cre/+) mouse can be used to generate an atrium-specific gene-targeting mutant, and it is based on the Cre-loxP system. In the present study, we used adult Sln(Cre/+) mice atria and analyzed the effects of heterozygous SLN deletion by Cre knockin before use as the gene targeting mouse. Both SLN mRNA and protein levels were decreased in Sln(Cre/+) mouse atria, but there were no morphological, physiological, or molecular biological abnormalities. The properties of contractility and Ca(2+) handling were similar to wild-type (WT) mice, and expression levels of several stress markers and sarcoplasmic reticulum-related protein levels were not different between Sln(Cre/+) and WT mice. Moreover, there was no significant difference in sarco(endo)plasmic reticulum Ca(2+)-ATPase activity between the two groups. We showed that Sln(Cre/+) mice were not significantly different from WT mice in all aspects that were examined. The present study provides basic characteristics of Sln(Cre/+) mice and possibly information on the usefulness of Sln(Cre/+) mice as an atrium-specific gene-targeting model.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-15, ascites fluid