Merck
Luteinizing Hormone and GATA4 Action in the Adrenocortical Tumorigenesis of Gonadectomized Female Mice.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-05)
Milena Doroszko, Marcin Chrusciel, Joanna Stelmaszewska, Tomasz Slezak, Adolfo Rivero-Muller, Artur Padzik, Slawomir Anisimowicz, Slawomir Wolczynski, Ilpo Huhtaniemi, Jorma Toppari, Nafis A Rahman
RESUMEN
Physiological role of luteinizing hormone (LH) and its receptor (LHCGR) in adrenal remains unknown. In inhibin-α/Simian Virus 40 T antigen (SV40Tag) (inhα/Tag) mice, gonadectomy-induced (OVX) elevated LH triggers the growth of transcription factor GATA4 (GATA4)-positive adrenocortical tumors in a hyperplasia-adenoma-adenocarcinoma sequence. We investigated the role of LHCGR in tumor induction, by crossbreeding inhα/Tag with Lhcgr knockout (LuRKO) mice. By knocking out Lhcgr and Gata4 in Cα1 adrenocortical cells (Lhcgr-ko, Gata4-ko) we tested their role in tumor progression. Adrenal tumors of OVX inhα/Tag mice develop from the hyperplastic cells localized in the topmost layer of zona fasciculata. OVX inhα/Tag/LuRKO only developed SV40Tag positive hyperplastic cells that were GATA4 negative, cleaved caspase-3 positive and did not progress into adenoma. In contrast to Lhcgr-ko, Gata4-ko Cα1 cells presented decreased proliferation, increased apoptosis, decreased expression of Inha, SV40Tag and Lhcgr tumor markers, as well as up-regulated adrenal- and down-regulated sex steroid gene expression. Both Gata4-ko and Lhcgr-ko Cα1 cells had decreased expression of steroidogenic genes resulting in decreased basal progesterone production. Our data indicate that LH/LHCGR signaling is critical for the adrenal cell reprogramming by GATA4 induction prompting adenoma formation and gonadal-like phenotype of the adrenocortical tumors in inhα/Tag mice.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
TWEEN® 20, viscous liquid
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
TWEEN® 20, viscous liquid
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad