Merck
Synthesis of Tetrahydrobiphenylene via Pd(0)-Catalyzed C(sp
Chemical & pharmaceutical bulletin (2017-12-05)
Chihiro Tsukano, Satoshi Suetsugu, Nobusuke Muto, Yoshiji Takemoto
RESUMEN
Tetrahydrobiphenylene consists of cyclobutene fused with benzene and cyclohexene rings. In this paper, a direct method for synthesizing tetrahydrobiphenylenes based on a palladium (Pd)(0)-catalyzed C(sp
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
o-Tolylmagnesium bromide solution, 2.0 M in diethyl ether