- Lysozyme oxidation by singlet molecular oxygen: Peptide characterization using [
Lysozyme oxidation by singlet molecular oxygen: Peptide characterization using [
Journal of mass spectrometry : JMS (2017-08-13)
Emerson Finco Marques, Marisa H G Medeiros, Paolo Di Mascio
PMID28801970
RESUMEN
Singlet molecular oxygen (