Merck
PGE
American journal of physiology. Renal physiology (2017-07-14)
Alexis A Gonzalez, Nicolas Salinas-Parra, Dan Leach, L Gabriel Navar, Minolfa C Prieto
RESUMEN
During the early phase of ANG II-dependent hypertension, tubular PGE
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
L-798106, ≥98% (HPLC)