Merck
  • Wogonoside alleviates inflammation induced by traumatic spinal cord injury by suppressing NF-κB and NLRP3 inflammasome activation.
Wogonoside alleviates inflammation induced by traumatic spinal cord injury by suppressing NF-κB and NLRP3 inflammasome activation.
Experimental and therapeutic medicine (2017-10-03)
Yonglin Zhu, Hanzhong Zhu, Zhaojie Wang, Fengguang Gao, Jingsheng Wang, Wenqiang Zhang
RESUMEN
Wogonoside possesses anti-oxidative, anti-inflammatory, anti-allergy and anti-tumor properties. The aim of the present study was to evaluate whether wogonoside alleviates spinal cord injury (SCI)-induced inflammation via nuclear factor (NF)-κB and nucleotide-binding oligomerization domain-like receptor family pyrin domain containing 3 (NLRP3) inflammasome activation. Sprague-Dawley rats were positioned in the jaws of a calibrated aneurysm clip with a closing pressure of 55 g. The jaws were placed on the dorsal and ventral surfaces of the spinal cord and left in place for 1 min. SCI rats were treated with 12, 25 and 50 mg/kg wogonoside. Following this, the locomotor function was assessed using the Basso Beattie Bresnahan scale. The water content of the spinal cord was measured, tumor necrosis factor-α (TNF-α), interleukin (IL)-1β and IL-6 levels were assessed and western blot analysis was performed to evaluate the expressions of NF-κB and NLRP3. Wogonoside was demonstrated to significantly ameliorate the SCI-induced reduction in Basso Beattie Bresnahan score (P<0.01) and significantly reduce the water content of the spinal cord in rats with SCI-induced inflammation (P<0.01). Results also indicated that treatment with wogonoside significantly reduced the levels of IL-1β, TNF-α and IL-6 in rats with SCI-induced inflammation (P<0.01), potentially via the phosphorylation of NF-κB inhibitor α. Furthermore, treatment with wogonoside inhibited the expressions of toll-like receptor 4, NLRP3 and caspase-1 protein in SCI model rats (P<0.01). In conclusion, the results of the present study suggest that wogonoside alleviates SCI-induced inflammation by suppressing NF-κB and NLRP3 inflammasome activation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Wogonoside, ≥95% (LC/MS-ELSD)