Merck
Interferon γ is a STAT1-dependent direct inducer of BCL6 expression in imatinib-treated chronic myeloid leukemia cells.
Oncogene (2017-04-04)
H S Madapura, N Nagy, D Ujvari, T Kallas, M C L Kröhnke, S Amu, M Björkholm, L Stenke, P K Mandal, J S McMurray, M Keszei, L S Westerberg, H Cheng, F Xue, G Klein, E Klein, D Salamon
RESUMEN
B-cell CLL/lymphoma 6 (BCL6) exerts oncogenic effects in several human hematopoietic malignancies including chronic myeloid leukemia (CML), where BCL6 expression was shown to be essential for CML stem cell survival and self-renewal during imatinib mesylate (IM) treatment. As several lines of evidence suggest that interferon γ (IFNγ) production in CML patients might have a central role in the response to tyrosine kinase inhibitor (TKI) therapy, we analyzed if IFNγ modulates BCL6 expression in CML cells. Although separate IFNγ or IM treatment only slightly upregulated BCL6 expression, combined treatment induced remarkable BCL6 upregulation in CML lines and primary human CD34+ CML stem cells. We proved that during combined treatment, inhibition of constitutive signal transducer and activator of transcription (STAT) 5 activation by IM allowed the specific enhancement of the STAT1 dependent, direct upregulation of BCL6 by IFNγ in CML cells. By using colony-forming assay, we found that IFNγ enhanced the ex vivo colony or cluster-forming capacity of human CML stem cells in the absence or presence of IM, respectively. Furthermore, inhibition of the transcriptional repressor function of BCL6 in the presence of IM and IFNγ almost completely blocked the cluster formation of human CML stem cells. On the other hand, by using small interfering RNA knockdown of BCL6, we demonstrated that in an IM-treated CML line the antiapoptotic effect of IFNγ was independent of BCL6 upregulation. We found that IFNγ also upregulated several antiapoptotic members of the BCL2 and BIRC gene families in CML cells, including the long isoform of MCL1, which proved to be essential for the antiapoptotic effect of IFNγ in an IM-treated CML line. Our results suggest that combination of TKIs with BCL6 and MCL1 inhibitors may potentially lead to the complete eradication of CML stem cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
FX1, ≥98% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad