Merck
Hypercoagulability progresses to hypocoagulability during evolution of acetaminophen-induced acute liver injury in pigs.
Scientific reports (2017-08-26)
Karla Chui Luan Lee, Luisa Baker, Susan Mallett, Anne Riddell, Pratima Chowdary, Hatim Alibhai, Yu-Mei Chang, Simon Priestnall, Giacomo Stanzani, Nathan Davies, Rajeshwar Mookerjee, Rajiv Jalan, Banwari Agarwal
RESUMEN
Increases in prothrombin time (PT) and international normalised ratio (INR) characterise acute liver injury (ALI) and failure (ALF), yet a wide heterogeneity in clotting abnormalities exists. This study defines evolution of coagulopathy in 10 pigs with acetaminophen (APAP)-induced ALI compared to 3 Controls. APAP administration began at 0 h and continued to 'ALF', defined as INR >3. In APAP pigs, INR was 1.05 ± 0.02 at 0 h, 2.15 ± 0.43 at 16 h and > 3 at 18 ± 1 h. At 12 h thromboelastography (TEG) demonstrated increased clot formation rate, associated with portal vein platelet aggregates and reductions in protein C, protein S, antithrombin and A Disintegrin and Metalloprotease with Thrombospondin type 1 repeats-13 (ADAMTS-13) to 60%, 24%, 47% and 32% normal respectively. At 18 ± 1 h, INR > 3 was associated with: hypocoagulable TEG profile with heparin-like effect; falls in thrombin generation, Factor V and Factor VIII to 52%, 19% and 17% normal respectively; further decline in anticoagulants; thrombocytopenia; neutrophilia and endotoxemia. Multivariate analysis, found that ADAMTS-13 was an independent predictor of a hypercoagulable TEG profile and platelet count, endotoxin, Protein C and fibrinogen were independent predictors of a hypocoagulable TEG profile. INR remained normal in Controls. Dynamic changes in coagulation occur with progression of ALI: a pro-thrombotic state progresses to hypocoagulability.