- First Bispecific Inhibitors of the Epidermal Growth Factor Receptor Kinase and the NF-κB Activity As Novel Anticancer Agents.
First Bispecific Inhibitors of the Epidermal Growth Factor Receptor Kinase and the NF-κB Activity As Novel Anticancer Agents.
Journal of medicinal chemistry (2017-03-16)
Mostafa M Hamed, Sarah S Darwish, Jennifer Herrmann, Ashraf H Abadi, Matthias Engel
PMID28291344
RESUMEN
The activation of the NF-κB transcription factor is a major adaptive response induced upon treatment with EGFR kinase inhibitors, leading to the emergence of resistance in nonsmall cell lung cancer and other tumor types. To suppress this survival mechanism, we developed new thiourea quinazoline derivatives that are dual inhibitors of both EGFR kinase and the NF-κB activity. Optimization of the hit compound, identified in a NF-κB reporter gene assay, led to compound 9b, exhibiting a cellular IC
MATERIALES
Referencia del producto
Marca
Descripción del producto
Corning® cell scrapers, blade L 1.8 cm, handle L 25 cm, sterile, case of 100