Merck
Activation of liver stromal cells is associated with male-biased liver tumor initiation in xmrk and Myc transgenic zebrafish.
Scientific reports (2017-09-06)
Qiqi Yang, Chuan Yan, Zhiyuan Gong
RESUMEN
Hepatocellular carcinoma (HCC) is more prevalent in men than in women. Previously we have found that some stromal cells, including hepatic stellate cells (HSCs), neutrophils and macrophages, play crucial roles in promoting sex disparity in kras