- A hybrid material as a sorbent phase for the disposable pipette extraction technique enhances efficiency in the determination of phenolic endocrine-disrupting compounds.
A hybrid material as a sorbent phase for the disposable pipette extraction technique enhances efficiency in the determination of phenolic endocrine-disrupting compounds.
Journal of chromatography. A (2017-07-25)
Gabriela Corazza, Josias Merib, Hérica A Magosso, Otávio R Bittencourt, Eduardo Carasek
PMID28734604
RESUMEN
In this study, the hybrid material 3-n-propyl(3-methylpyridinium) silsesquioxane chloride (Si3Py
MATERIALES
Número de producto