Merck
Identification of a rare LAMB4 variant associated with familial diverticulitis through exome sequencing.
Human molecular genetics (2017-06-09)
Joel L Coble, Kathryn E Sheldon, Feng Yue, Tarik J Salameh, Leonard R Harris, Sue Deiling, Francesca M Ruggiero, Melanie A Eshelman, Gregory S Yochum, Walter A Koltun, Glenn S Gerhard, James R Broach
RESUMEN
Diverticulitis is a chronic disease of the colon in which diverticuli, or outpouching through the colonic wall, become inflamed. Although recent observations suggest that genetic factors may play a significant role in diverticulitis, few genes have yet been implicated in disease pathogenesis and familial cases are uncommon. Here, we report results of whole exome sequencing performed on members from a single multi-generational family with early onset diverticulitis in order to identify a genetic component of the disease. We identified a rare single nucleotide variant in the laminin β 4 gene (LAMB4) that segregated with disease in a dominant pattern and causes a damaging missense substitution (D435N). Targeted sequencing of LAMB4 in 148 non-familial and unrelated sporadic diverticulitis patients identified two additional rare variants in the gene. Immunohistochemistry indicated that LAMB4 localizes to the myenteric plexus of colonic tissue and patients harboring LAMB4 variants exhibited reduced LAMB4 protein levels relative to controls. Laminins are constituents of the extracellular matrix and play a major role in regulating the development and function of the enteric nervous system. Reduced LAMB4 levels may therefore alter innervation and morphology of the enteric nervous system, which may contribute to colonic dysmotility associated with diverticulitis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Citrate Buffer, pH 6.0, 10×, Antigen Retriever, for immunohistochemistry
En este momento no podemos mostrarle ni los precios ni la disponibilidad