Merck
Hampered cumulus expansion of porcine cumulus-oocyte complexes by excessive presence of alpha
Animal science journal = Nihon chikusan Gakkaiho (2017-01-27)
Ruth Appeltant, Josine Beek, Dominiek Maes, Jo Bijttebier, Katleen Van Steendam, Hans Nauwynck, Ann Van Soom
RESUMEN
In vitro maturation (IVM) in serum causes hampered expansion of porcine cumulus-oocyte complexes (COCs) due to excessive alpha
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Tissue Inhibitor of Metalloproteinase-3 human, recombinant, expressed in NSO cells, >95% (SDS-PAGE)