Merck
Enzymatic synthesis of novel corylifol A glucosides via a UDP-glycosyltransferase.
Carbohydrate research (2017-05-22)
Nan Li, Jian Miao, Jing Li, Yu-Ru Zhao, Hong-Mei Li, Yi-Qun Dai, Qiang Huo, Cheng-Zhu Wu, Tao Ma
RESUMEN
Corylifol A, a member of the isoflavone subclass of isoflavonoids, has long been considered to have various biological activities. Here, we sought to synthesize corylifol A glucosides by the in vitro glucosylation reaction using the UDP-glycosyltransferase YjiC from Bacillus licheniformis DSM 13, and obtained two novel glucosides: corylifol A-4',7-di-O-beta-d-glucopyranoside (1) and corylifol A-4'-O-beta-d-glucopyranoside (2). To improve the yield of the products, the reaction time, concentration of UDP-glucose, and pH of the buffer were optimized. The Michaelis constant (K
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Uridine 5′-diphosphoglucose disodium salt, ≥98.0% (HPLC)