Merck
  • Hydroxyl radical generation by dissociation of water molecules during 1.65 MHz frequency ultrasound irradiation under aerobic conditions.
Hydroxyl radical generation by dissociation of water molecules during 1.65 MHz frequency ultrasound irradiation under aerobic conditions.
Biochemical and biophysical research communications (2017-01-04)
Akimitsu Miyaji, Masahiro Kohno, Yoshihiro Inoue, Toshihide Baba
RESUMEN
The dissociation of water molecules by ultrasound irradiation under aerobic conditions was demonstrated experimentally. To be able to detect the dissociation of water molecules, we performed the ultrasound irradiation of
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1-Hydroxypiperidine, ≥96.0% (GC)