Merck
  • Ultraviolet A eye irradiation ameliorates colon carcinoma induced by azoxymethane and dextran sodium sulfate through β-endorphin and methionine-enkephalin.
Ultraviolet A eye irradiation ameliorates colon carcinoma induced by azoxymethane and dextran sodium sulfate through β-endorphin and methionine-enkephalin.
Photodermatology, photoimmunology & photomedicine (2017-01-01)
Keiichi Hiramoto, Satoshi Yokoyama, Yurika Yamate
RESUMEN
We previously reported that ultraviolet (UV) A eye irradiation reduces the ulcerative colitis induced by dextran sodium sulfate (DSS). This study examined the effects of UVA on colon carcinoma induced by azoxymethane (AOM) and DSS. We irradiated the eyes of ICR mice with UVA at a dose of 110 kJ/m In mice treated with these drugs, the symptom of colon carcinoma was reduced by UVA eye irradiation. The levels of interleukin (IL)-6 and tumor necrosis factor (TNF)-α in the blood were increased in AOM + DSS-treated mice; however, those levels were reduced by UVA eye irradiation. The expression of β-endorphin, methionine-enkephalin (OGF), μ-opioid receptor, and opioid growth factor receptor (OGFR) of the colon was increased in the AOM + DSS-treated mice, and these levels were increased further following UVA eye irradiation. When β-endorphin inhibitor was administered, the ameliorative effect of UVA eye irradiation was reduced, and the effect of eye irradiation disappeared entirely following the administration of naltrexone (inhibitor of both opioid receptor and OGFR). These results suggested that UVA eye irradiation exerts major effects on AOM + DSS-induced colon carcinoma.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
JTC-801, ≥98% (HPLC)