Merck
A useful PET probe [
Nuclear medicine and biology (2016-11-12)
Kazunori Kawamura, Yoko Shimoda, Joji Yui, Yiding Zhang, Tomoteru Yamasaki, Hidekatsu Wakizaka, Akiko Hatori, Lin Xie, Katsushi Kumata, Masayuki Fujinaga, Masanao Ogawa, Yusuke Kurihara, Nobuki Nengaki, Ming-Rong Zhang
RESUMEN
A positron emission tomography (PET) probe with ultra-high specific radioactivity (SA) enables measuring high receptor specific binding in brain regions by avoiding mass effect of the PET probe itself. It has been reported that PET probe with ultra-high SA can detect small change caused by endogenous or exogenous ligand. Recently, Kealey et al. developed [ [ [ A PET study using [
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
BU224 hydrochloride, solid