Merck
  • N-Acylated chitosan bis(arylcarbamate)s: A class of promising chiral separation materials with powerful enantioseparation capability and high eluents tolerability.
N-Acylated chitosan bis(arylcarbamate)s: A class of promising chiral separation materials with powerful enantioseparation capability and high eluents tolerability.
Journal of chromatography. A (2016-11-20)
Sheng Tang, Jian-De Liu, Qin Bin, Ke-Qin Fu, Xiao-Chen Wang, Ying-Bin Luo, Shao-Hua Huang, Zheng-Wu Bai
RESUMEN
In order to comprehensively understand the influence of coordination of the substituent at 2-position with those at 3- and 6-positions on the properties of chitosan derivatives, a series of chitosan 3,6-bis(arylcarbamate)-2-(amide)s (CACAs) and the related chiral stationary phases (CSPs) were prepared and reported in the present study. Specifically, chitosan was N-acylated with carboxylic acid anhydrides, and then further derivatized with various aryl isocyanates to afford CACAs, from which a class of coated-type CSPs were prepared. When the substituent introduced on the acyl group at 2-position and those on the phenyl group of the carbamates at 3- and 6-positions were fittingly combined, these prepared CACAs based CSPs would exhibit powerful chiral recognition ability, further resulting in a class of promising chiral separation materials with excellent enantioseparation performance. Meanwhile, these newly developed materials with suitable molecular weight also bear a high tolerability towards organic solvents, even including pure tetrahydrofuran, thus broadening their application in enantiomeric separation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
p-Toluoyl chloride, 98%