Merck
  • Hydrated and Solvated Tin(II) Ions in Solution and the Solid State, and a Coordination Chemistry Overview of the d10 s2 Metal Ions.
Hydrated and Solvated Tin(II) Ions in Solution and the Solid State, and a Coordination Chemistry Overview of the d10 s2 Metal Ions.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-11-20)
Ingmar Persson, Paola D'Angelo, Daniel Lundberg
RESUMEN
The coordination chemistry of d10 s2 metal ions is strongly affected by an (at least partially) occupied d10 s2 metal ion-ligand atom antibonding orbital, which may cause a void in the coordination sphere due to repulsion between the electrons in the antibonding orbital on the metal ion and those on the ligands. The character of the formed d10 s2 metal ion-ligand atom bond plays an important role in the electron density in the antibonding orbital and thereby also in the coordination chemistry. The hydrated tin(II) ion, [Sn(H2 O)3 ]2+ , and the trihydroxidostannate ion, [Sn(OH)3 ]- , have very different mean Sn-O bond lengths (2.21 and 2.08 Å, respectively) and O-Sn-O angles (ca. 78 and 90°, respectively) both in the solid state and in solution. On increasing the covalency of the tin(II)-ligand bonds, the repulsion decreases and higher coordination numbers are obtained, as seen in the dimethylsulfoxide- and N,N-dimethylthioformamide-solvated tin(II) ions, both of which are five-coordinate with square-pyramidal structures.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N,N-Dimethylthioformamide, ≥98%