Merck
  • A highly regio- and stereoselective formation of bicyclo[4.2.0]oct-5-ene derivatives through thermal intramolecular [2 + 2] cycloaddition of allenes.
A highly regio- and stereoselective formation of bicyclo[4.2.0]oct-5-ene derivatives through thermal intramolecular [2 + 2] cycloaddition of allenes.
The Journal of organic chemistry (2007-05-11)
Hiroaki Ohno, Tsuyoshi Mizutani, Yoichi Kadoh, Akimasa Aso, Kumiko Miyamura, Nobutaka Fujii, Tetsuaki Tanaka
RESUMEN
Thermal [2 + 2] cycloaddition of allenes with an additional multiple bond is described. By simply heating the allenenes or allenynes having a three-atom tether in an appropriate solvent such as dioxane or DMF, the distal double bond of the allenic moiety regioselectively participates in the cycloaddition to form bicyclo[4.2.0]oct-5-ene derivatives in good to excellent yields. In all the reactions of allenenes, the olefin geometry was completely transferred to the cycloadducts. While the reaction of terminal allenes afforded bicyclic cyclobutane derivatives as a single isomer, the cycloaddition of some internal allenes with axial chirality yielded a diastereomeric mixture of cycloadducts. These results are in good accordance with the stepwise mechanism through a biradical intermediate with a coplanar allyl radical.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(Triphenylphosphoranylidene)acetonitrile, 97%