Merck
Density-dependent cooperative non-specific binding in solid-phase SELEX affinity selection.
Nucleic acids research (2013-06-06)
Abdullah Ozer, Brian S White, John T Lis, David Shalloway
RESUMEN
The non-specific binding of undesired ligands to a target is the primary factor limiting the enrichment of tight-binding ligands in affinity selection. Solution-phase non-specific affinity is determined by the free-energy of ligand binding to a single target. However, the solid-phase affinity might be higher if a ligand bound concurrently to multiple adjacent immobilized targets in a cooperative manner. Cooperativity could emerge in this case as a simple consequence of the relationship between the free energy of binding, localization entropy and the spatial distribution of the immobilized targets. We tested this hypothesis using a SELEX experimental design and found that non-specific RNA aptamer ligands can concurrently bind up to four bead-immobilized peptide targets, and that this can increase their effective binding affinity by two orders-of-magnitude. Binding curves were quantitatively explained by a new statistical mechanical model of density-dependent cooperative binding, which relates cooperative binding to both the target concentration and the target surface density on the immobilizing substrate. Target immobilization plays a key role in SELEX and other ligand enrichment methods, particularly in new multiplexed microfluidic purification devices, and these results have strong implications for optimizing their performance.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Activated Thiol–Sepharose 4B, lyophilized powder