Merck
  • Efficient catalytic enantioselective synthesis of unsaturated amines: preparation of small- and medium-ring cyclic amines through mo-catalyzed asymmetric ring-closing metathesis in the absence of solvent.
Efficient catalytic enantioselective synthesis of unsaturated amines: preparation of small- and medium-ring cyclic amines through mo-catalyzed asymmetric ring-closing metathesis in the absence of solvent.
Journal of the American Chemical Society (2002-06-13)
Sarah J Dolman, Elizabeth S Sattely, Amir H Hoveyda, Richard R Schrock
RESUMEN
The first catalytic asymmetric ring-closing metathesis method for the synthesis of N-containing heterocycles is reported; this is accomplished through Mo-catalyzed kinetic resolution or desymmetrization of unsaturated amines. Importantly, this catalytic asymmetric method delivers medium-ring unsaturated amines (including eight-membered rings) in high yield, with exceptional enantioselectivity and without the need for solvents. These enantioselective reactions can be effected by catalysts prepared in situ from commercially available reagents.