Merck
Molecular characterisation of long-acting insulin analogues in comparison with human insulin, IGF-1 and insulin X10.
PloS one (2012-05-17)
Bo F Hansen, Tine Glendorf, Anne C Hegelund, Anders Lundby, Anne Lützen, Rita Slaaby, Carsten Enggaard Stidsen
RESUMEN
There is controversy with respect to molecular characteristics of insulin analogues. We report a series of experiments forming a comprehensive characterisation of the long acting insulin analogues, glargine and detemir, in comparison with human insulin, IGF-1, and the super-mitogenic insulin, X10. We measured binding of ligands to membrane-bound and solubilised receptors, receptor activation and mitogenicity in a number of cell types. Detemir and glargine each displayed a balanced affinity for insulin receptor (IR) isoforms A and B. This was also true for X10, whereas IGF-1 had a higher affinity for IR-A than IR-B. X10 and glargine both exhibited a higher relative IGF-1R than IR binding affinity, whereas detemir displayed an IGF-1R:IR binding ratio of ≤ 1. Ligands with high relative IGF-1R affinity also had high affinity for IR/IGF-1R hybrid receptors. In general, the relative binding affinities of the analogues were reflected in their ability to phosphorylate the IR and IGF-1R. Detailed analysis revealed that X10, in contrast to the other ligands, seemed to evoke a preferential phosphorylation of juxtamembrane and kinase domain phosphorylation sites of the IR. Sustained phosphorylation was only observed from the IR after stimulation with X10, and after stimulation with IGF-1 from the IGF-1R. Both X10 and glargine showed an increased mitogenic potency compared to human insulin in cells expressing many IGF-1Rs, whereas only X10 showed increased mitogenicity in cells expressing many IRs. Detailed analysis of receptor binding, activation and in vitro mitogenicity indicated no molecular safety concern with detemir.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Lectin from Triticum vulgaris (wheat), agarose conjugate, saline suspension
En este momento no podemos mostrarle ni los precios ni la disponibilidad