Merck
Activation of Estrogen Receptor by Bavachin from Psoralea corylifolia.
Biomolecules & therapeutics (2012-03-01)
Joonwoo Park, Do Hee Kim, Hye-Na Ahn, Yun Seon Song, Young Joo Lee, Jae-Ha Ryu
RESUMEN
In this study, we examined the estrogenic activity of bavachin, a component of Psoralea corylifolia that has been used as a traditional medicine in Asia. Bavachin was purified from ethanolic extract of Psoralea corylifolia and characterized its estrogenic activity by ligand binding, reporter gene activation, and endogenous estrogen receptor (ER) target gene regulation. Bavachin showed ER ligand binding activity in competitive displacement of [(3)H] E2 from recombinant ER. The estrogenic activity of bavachin was characterized in a transient transfection system using ERα or ERβ and estrogen-responsive luciferase plasmids in CV-1 cells with an EC50 of 320 nM and 680 nM, respectively. Bavachin increased the mRNA levels of estrogen-responsive genes such as pS2 and PR, and decreased the protein level of ERα by proteasomal pathway. However, bavachin failed to activate the androgen receptor in CV-1 cells transiently transfected with the corresponding receptor and hormone responsive reporter plasmid. These data indicate that bavachin acts as a weak phytoestrogen by binding and activating the ER.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Bavachin, from Psoralea coryfolia, ≥97% (HPLC)