Merck
  • Oxidative stress induces the acquisition of cancer stem-like phenotype in breast cancer detectable by using a Sox2 regulatory region-2 (SRR2) reporter.
Oxidative stress induces the acquisition of cancer stem-like phenotype in breast cancer detectable by using a Sox2 regulatory region-2 (SRR2) reporter.
Oncotarget (2015-12-20)
Keshav Gopal, Nidhi Gupta, Haifeng Zhang, Abdulraheem Alshareef, Hind Alqahtani, Gilbert Bigras, Jamie Lewis, Donna Douglas, Norman Kneteman, Afsaneh Lavasanifar, Raymond Lai
RESUMEN
We have previously identified a novel intra-tumoral dichotomy in breast cancer based on the differential responsiveness to a Sox2 reporter (SRR2), with cells responsive to SRR2 (RR) being more stem-like than unresponsive cells (RU). Here, we report that RR cells derived from MCF7 and ZR751 displayed a higher tolerance to oxidative stress than their RU counterparts, supporting the concept that the RR phenotype correlates with cancer stemness. Sox2 is directly implicated in this differential H2O2 tolerance, since siRNA knockdown of Sox2 in RR cells leveled this difference. Interestingly, H2O2 converted a proportion of RU cells into RR cells, as evidenced by their expression of luciferase and GFP, markers of SRR2 activity. Compared to RU cells, converted RR cells showed a significant increase in mammosphere formation and tolerance to H2O2. Converted RR cells also adopted the biochemical features of RR cells, as evidenced by their substantial increase in Sox2-SRR2 binding and the expression of 3 signature genes of RR cells (CD133, GPR49 and MUC15). Lastly, the H2O2-induced RU/RR conversion was detectable in a SCID mouse xenograft model and primary tumor cells. To conclude, the H2O2-induced RU/RR conversion has provided a novel model to study the acquisition of cancer stemness and plasticity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human SOX2