Merck
  • Transient transfection of macrophage migration inhibitory factor small interfering RNA disrupts the biological behavior of oral squamous carcinoma cells.
Transient transfection of macrophage migration inhibitory factor small interfering RNA disrupts the biological behavior of oral squamous carcinoma cells.
Molecular medicine reports (2015-11-10)
Jie Zeng, Jingjing Quan, Xuefeng Xia
RESUMEN
Macrophage migration inhibitory factor (MIF) is closely associated with tumorigenesis. The present study aimed to investigate the effects of MIF on the proliferation, migration and colony formation of oral squamous cell carcinoma (OSCC), and to quantify the protein expression levels of MIF in OSCC tissue samples. Firstly, small interfering (si)RNA was used to knock down the gene expression of MIF in Tca8113, HN5 and SCC25 OSCC cells. Secondly, proliferation, migration and colony formation of the OSCC cells were determined by MTT, transmigration and colony formation assays, respectively. Western blotting was performed to detect changes in the protein expression levels of the epithelial mesenchymal transition markers, Twist‑related protein 1 (Twist1), matrix metalloproteinase (MMP)‑2 and MMP‑9. Finally, immunohistochemistry was used to examine the protein expression of MIF in OSCC tissue samples. The results demonstrated that siRNA against MIF significantly downregulated the expression levels of MIF in all OSCC cells, and decreased their proliferation and migration ability. Colony formation ability was also inhibited in the OSCC cells following transfection with MIF siRNA. Furthermore, western blotting demonstrated that the protein expression of Twist1 was decreased similarly to those of MIF. The protein expression of MMP‑2 revealed no change, whereas that of MMP‑9 decreased. The protein expression of MIF was detected in OSCC tissue samples with staining predominantly located in the cell membrane and cytoplasm. The present study demonstrated that MIF may be important in the pathogenesis and progression of OSCC, and indicated its potential therapeutic value.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human MIF
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mif