Merck
PINK1 heterozygous mutations induce subtle alterations in dopamine-dependent synaptic plasticity.
Movement disorders : official journal of the Movement Disorder Society (2013-10-30)
Graziella Madeo, Tommaso Schirinzi, Giuseppina Martella, E Claudio Latagliata, Francesca Puglisi, Jie Shen, Enza Maria Valente, Mauro Federici, Nicola B Mercuri, Stefano Puglisi-Allegra, Paola Bonsi, Antonio Pisani
RESUMEN
Homozygous or compound heterozygous mutations in the phosphatase and tensin homolog-induced putative kinase 1 (PINK1) gene are causative of autosomal recessive, early onset Parkinson's disease. Single heterozygous mutations have been detected repeatedly both in a subset of patients and in unaffected individuals, and the significance of these mutations has long been debated. Several neurophysiological studies from non-manifesting PINK1 heterozygotes have demonstrated the existence of neural plasticity abnormalities, indicating the presence of specific endophenotypic traits in the heterozygous state. We performed a functional analysis of corticostriatal synaptic plasticity in heterozygous PINK1 knockout (PINK1(+/-) ) mice using a multidisciplinary approach and observed that, despite normal motor behavior, repetitive activation of cortical inputs to striatal neurons failed to induce long-term potentiation (LTP), whereas long-term depression was normal. Although nigral dopaminergic neurons exhibited normal morphological and electrophysiological properties with normal responses to dopamine receptor activation, a significantly lower dopamine release was measured in the striatum of PINK1(+/-) mice compared with control mice, suggesting that a decrease in stimulus-evoked dopamine overflow acts as a major determinant for the LTP deficit. Accordingly, pharmacological agents capable of increasing the availability of dopamine in the synaptic cleft restored normal LTP in heterozygous mice. Moreover, monoamine oxidase B inhibitors rescued physiological LTP and normal dopamine release. Our results provide novel evidence for striatal plasticity abnormalities, even in the heterozygous disease state. These alterations might be considered an endophenotype to this monogenic form of Parkinson's disease and a valid tool with which to characterize early disease stage and design possible disease-modifying therapies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Extract-N-Amp Tissue PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
Extract-N-Amp Tissue PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications