Merck
Transcriptional activation of p21(WAF1/CIP1) by apicidin, a novel histone deacetylase inhibitor.
Biochemical and biophysical research communications (2001-03-10)
J S Kim, S Lee, T Lee, Y W Lee, J B Trepel
RESUMEN
Apicidin [cyclo(N-O-methyl-L-tryptophanyl-L-isoleucinyl-D-pipecolinyl-l-2-amino-8-oxodecanoyl)], a novel histone deacetylase inhibitor, has been identified as an antiprotozoal and antiproliferative agent. In this study, we show apicidin induces transcriptional activation of p21(WAF1/CIP1) (p21) in human prostate carcinoma cells. Apicidin induces expression of p21 protein and mRNA and activation of p21 promoter-luciferase reporter constructs. Apicidin causes an accumulation of acetylated histones H3 and H4 in total cellular chromatin. Chromatin immunoprecipitation shows p21 promoter DNA is associated with hyperacetylated histones H3 and H4 after treatment with apicidin. Therefore, the data here demonstrate that apicidin activates p21 transcription associated with the acetylation of histones H3 and H4.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial