Merck
Hydroxyflavanone inhibits gastric carcinoma MGC-803 cell proliferation.
International journal of clinical and experimental medicine (2015-12-03)
Haiyan Zhang, Zhuo Zhan, Mingfu Cui, Yongjian Gao, Dayu Wang, Ye Feng
RESUMEN
Gastric carcinoma (GC) is the most common primary malignancy of the digestive tract, with increasing incidence in many countries. 3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay was used to assess inhibition of HepG2 cell proliferation by 2'-hydroxyflavanone. The STAT3 pathway was performed. 2'-hydroxyflavanone reduced inhibitory effects on MGC-803 cell proliferation. 2'-hydroxyflavanone exhibited the highest inhibition rate. Treatment of MGC-803 cells with 400, 200, and 100 μg/ml 2'-hydroxyflavanone resulted in 88.9±0.7%, 81.2±0.5%, 68.4±0.5% decrease in cell viability, respectively, indicating an IC50 of 9.3 μg/ml. The 100 μg/ml 2'-hydroxyflavanone can significantly inhibit the STAT3 pathway activation. 2'-hydroxyflavanone inhibits MGC-803 cell proliferation by inhibiting STAT3 pathway activation. This extract is therefore a potential drug candidate for treatment of liver cancer.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2′-Hydroxyflavanone, ≥98%