Merck
Clinical significance of the uPA system in gastric cancer with peritoneal metastasis.
European journal of medical research (2013-08-30)
Youcheng Ding, Hui Zhang, Mingan Zhong, Zhuqing Zhou, Zhixiang Zhuang, Hua Yin, Xujing Wang, Zhenggang Zhu
RESUMEN
It has been demonstrated that urokinase-type plasminogen activator (uPA) is involved in tumor cell metastasis by degrading the extracellular matrix. However, there is little direct evidence of clinical uPA system expression in peritoneal metastatic tissues of gastric cancer. The objective of this study was to investigate uPA system expression in peritoneal tissues of peritoneal and nonperitoneal metastasis patients, and to explore the diagnostic value of the uPA system. Expressions of uPA, uPAR, and PAI-1 were measured by semi-quantitative RT-PCR and ELISA. uPA activity was detected using a uPA activity kit. There was no significant difference in uPA, uPAR, and PAI-1 expression in two types of peritoneal tissue in seven patients with peritoneal metastasis. However, uPA, uPAR, and PAI-1 expressions in peritoneal metastatic lesions were significantly higher than those in normal peritoneal tissues of 24 nonperitoneal metastasis patients (P <0.05). Moreover, no statistical discrepancy of uPA activity was observed in various different tissues. The expression of the uPA system positively correlates with peritoneal metastasis of gastric cancer. This expression difference in peritoneal or nonperitoneal metastasis patients may provide a reference for diagnosis of peritoneal metastasis.