Merck
Lysosomal cysteine proteases: facts and opportunities.
The EMBO journal (2001-09-05)
V Turk, B Turk, D Turk
RESUMEN
From their discovery in the first half of the 20th century, lysosomal cysteine proteases have come a long way: from being the enzymes non-selectively degrading proteins in lysosomes to being those responsible for a number of important cellular processes. Some of the features and roles of their structures, specificity, regulation and physiology are discussed.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cathepsin L Inhibitor