Merck
Protective efficacy of CAP18106-138-immunoglobulin G in sepsis.
The Journal of infectious diseases (2003-11-01)
H Shaw Warren, Robina Matyal, Jennifer E Allaire, David Yarmush, Paul Loiselle, Judith Hellman, Barbara G Paton, Mitchell P Fink
RESUMEN
Naturally present antibacterial proteins play an important role in innate host defense. A synthetic peptide mimicking the C-terminal lipopolysaccharide (LPS)-binding domain of rabbit cathelicidin CAP18 was coupled to immunoglobulin (Ig) G to create CAP18(106-138)-IgG, a construct that, in concentrations equimolar to those of peptide alone, binds and neutralizes LPS and kills multiple gram-negative bacterial strains. The protective efficacy of CAP18(106-138)-IgG was evaluated in a model of cecal ligation and puncture in mice. A single intravenous administration of 20 mg/kg CAP18(106-138)-IgG protected against mortality, compared with sham-coupled IgG (P<.03). There was no protection offered by administration of equimolar peptide alone (P=.96). There was a trend toward protection in C3H/HeJ mice that are minimally sensitive to LPS (P=.06), suggesting that direct detoxification of LPS was not the only mechanism of protection. Chemical or genetic coupling of antimicrobial peptides to IgG may be a means of using these peptides to treat infections.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
SMPT (4-succinimidyloxycarbonyl-alpha-methyl-alpha(2-pyridyldithio)toluene)