Merck
Phosphoramidon blocks big-endothelin-1 but not endothelin-1 enhancement of vascular permeability in the rat.
British journal of pharmacology (1992-12-01)
S Lehoux, G E Plante, M G Sirois, P Sirois, P D'Orléans-Juste
RESUMEN
1. Changes in vascular permeability following intravenous injections of human big-endothelin-1 (big-ET-1) and endothelin-1 (ET-1) were measured by extravasation of Evans blue dye (EB, 20 mg kg-1) in selected tissues. 2. A low dose of big-ET-1 (40 pmol kg-1) failed to alter vascular permeability but a dose of 400 pmol kg-1 increased EB extravasation in the trachea, upper and lower bronchi, and lung parenchyma by 55 to 69% (P < 0.05). Vascular permeability was also enhanced in the liver, spleen, kidney, heart, and diaphragm by 20, 14, 41, 25, and 67%, respectively (P < 0.05). 3. Upon injection of ET-1 (400 pmol kg-1), EB extravasation increased in the upper and lower bronchi, lung parenchyma, liver, pancreas, kidney, heart, and diaphragm. 4. Administration of ET-1 and big-ET-1 was not associated with significant systemic responses. 5. Pretreatment with phosphoramidon (PA) blocked the response to big-ET-1 in all tissues examined but this inhibitor failed to alter the response to ET-1. 6. We conclude from these results that the dose-dependent increase in vascular permeability induced by big-ET-1 in various tissues follows its conversion to ET-1 by the endothelin converting enzyme, a PA-sensitive process.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Big Endothelin 39 rat, ≥97% (HPLC)