MilliporeSigma
Myxoinflammatory fibroblastic sarcoma: a clinicopathologic analysis of 104 cases, with emphasis on predictors of outcome.
The American journal of surgical pathology (2013-10-15)
William B Laskin, John F Fetsch, Markku Miettinen
RESUMEN
In this study, we examine the clinicopathologic features of 104 cases of myxoinflammatory fibroblastic sarcoma (MIFS), a low-grade, inflammatory fibromyxoid tumor with a predilection to distal extremity soft tissue, and attempt to identify factors predictive of aggressive behavior. The study cohort consisted of 49 male and 55 female patients ranging in age from 17 to 83 (mean, 42; median, 39) years. The tumor arose primarily on the dorsal aspect of the distal extremities as a solitary and usually painless mass. Tumors ranged in size from 0.5 to 15 (mean, 3.2; median; 2.4) cm. Microscopically, tumors consisted of variably cellular and inflamed fibromyxoid tissue growing as a lobulated mass or as multiple nodules within subcutaneous tissue or along tendinofascial planes. Tumor cells ranged from plump spindled to more epithelioid cells with enlarged, vesicular nuclei. Characteristic of the process was a strikingly bizarre cell with an inclusion body-like nucleolus (85% of cases) and/or a smudgy hyperchromatic nucleus (51%) present in all but 7 cases. The mitotic rate per 50 high-power field ranged from 0 to 13 (mean, 2,9; median, 2) mitoses. Twenty-two tumors demonstrated 1 or more of the following atypical features: (1) foci with complex sarcoma-like vasculature; (2) hypercellular areas; and (3) increased mitotic activity or atypical mitotic figures. Immunohistochemically, tumor cells demonstrated immunoreactivity for vimentin (100%), D2-40 (86%), CD34 (50%), keratin(s) (33%), CD68 (27%), actin(s) (26%), desmin (9%), S-100 protein (7%), and epithelial membrane antigen (6%). Thirty of 59 patients (51%) with follow-up data suffered (at least) 1 local recurrence, and 1 patient developed metastatic disease after multiple local recurrences. Completeness of initial surgical excision was the only clinicopathologic parameter that statistically correlated with a lower incidence of recurrence (P=0.004). Histologically atypical MIFS recurred more often than conventional tumors (67% vs. 47%), but the difference was not statistically significant (P=0.35). Our study shows that histologic features often associated with more aggressive sarcomas do not substantially impact the morbidity of MIFS, and complete surgical excision provides the best chance for disease-free survival.