Merck
Factor VIIa binding to endothelial cell protein C receptor protects vascular barrier integrity in vivo.
Journal of thrombosis and haemostasis : JTH (2014-07-01)
J Sundaram, S Keshava, R Gopalakrishnan, C T Esmon, U R Pendurthi, L V M Rao
RESUMEN
Recent studies have shown that factor VIIa binds to endothelial cell protein C receptor(EPCR), a cellular receptor for protein C and activated protein C. At present, the physiologic significance of FVIIa interaction with EPCR in vivo remains unclear. To investigate whether exogenously administered FVIIa, by binding to EPCR, induces a barrier protective effect in vivo. Lipopolysaccharide(LPS)-induced vascular leakage in the lung and kidney,and vascular endothelial growth factor (VEGF)-induced vascular leakage in the skin, were used to evaluate the FVIIa-induced barrier protective effect. Wild-type, EPCR-deficient, EPCR-overexpressing and hemophilia A mice were used in the studies. Administration ofFVIIa reduced LPS-induced vascular leakage in the lung and kidney; the FVIIa-induced barrier protective effect was attenuated in EPCR-deficient mice. The extent of VEGF-induced vascular leakage in the skin was highly dependent on EPCR expression levels. Therapeutic concentrations of FVIIa attenuated VEGF-induced vascular leakage in control mice but not in EPCR-deficient mice.Blockade of FVIIa binding to EPCR with a blocking mAb completely attenuated the FVIIa-induced barrier protective effect. Similarly, administration of protease activated receptor 1 antagonist blocked the FVIIa induced barrier protective effect. Hemophilic mice showed increased vascular permeability, and administration of therapeutic concentrations of FVIIa improved barrier integrity in these mice. This is the first study to demonstrate that FVIIa binding to EPCR leads to a barrier protective effect in vivo. This finding may have clinical relevance, as it indicates additional advantages of using FVIIa in treating hemophilic patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Vascular Endothelial Growth Factor from mouse, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture